Drukuj
Kolumny kapilarne, OPTIMA® 5 Amine
Dostawca: MACHEREY-NAGEL
726353.30 726354.30 726355.25 726356.30 726357.30 726358.30 726359.30 726360.30 726361.10
MANA726353.30EA 3480 PLN
MANA726353.30 MANA726354.30 MANA726355.25 MANA726356.30 MANA726357.30 MANA726358.30 MANA726359.30 MANA726360.30 MANA726361.10
Kolumny kapilarne, OPTIMA® 5 Amine
Kolumny chromatograficzne
Special GC phase for the analysis of amines.
  • Especially deactivated for the analysis of polyfunctional amines such as ethanol­amines, amino-functionalized diols and similar compounds
  • Improved linearity for analyses of active components at trace levels
  • Max. temperature for isothermal operation 300 °C
  • Max. temperature for short isotherms in a temperature program 320 °C
USP code: G27 / G36.