Drukuj
Rurki do rozprowadzania gazów
Dostawca: SciLabware
3830/00M 3830/02M 3830/04M 3830/06M
BIBB3830/00MEA 621 PLN
BIBB3830/00M BIBB3830/02M BIBB3830/04M BIBB3830/06M
Rurki do rozprowadzania gazów
Rurki sorpcyjne
These Pyrex® borosilicate glass tubes for the dispersion of gas in liquids with sintered thimble allow for the choice of porosities to achieve optimum gas bubble size.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion with relatively high resistance to temperature changes