Drukuj
Kolumny kapilarne, OPTIMA® 1 MS
Dostawca: MACHEREY-NAGEL
726201.25 726201.50 726202.30 726202.60 726203.12 726205.15 726205.30 726205.60
MANA726201.25EA 3270 PLN
MANA726201.25 MANA726201.50 MANA726202.30 MANA726202.60 MANA726203.12 MANA726205.15 MANA726205.30 MANA726205.60
Kolumny kapilarne, OPTIMA® 1 MS
Kolumny chromatograficzne
Nonpolar GC phase with low bleeding.
  • Suited for GC/MS and ECD applications and general analyses at trace level
  • Maximum temperature for isothermal operation 340 °C
  • Maximum temperature for short isotherms in a temperature program 360 °C
100% dimethylpolysiloxane phase.
USP code: G1/G2/G38.