Drukuj
Filtry odśrodkowe, membrana mikroporowata Ultrafree®-MC
Dostawca: MILLIPORE
UFC30DV00 UFC30HVNB UFC30VV00 UFC30GVNB UFC30HV00 UFC30GV00 UFC30VV25 UFC30GV25 UFC30DV25 UFC30HV25 UFC30GV0S UFC30DV0S UFC30SV00 UFC30HVIS
MILYUFC30DV00EA 1980 PLN
MILYUFC30DV00 MILYUFC30HVNB MILYUFC30VV00 MILYUFC30GVNB MILYUFC30HV00 MILYUFC30GV00 MILYUFC30VV25 MILYUFC30GV25 MILYUFC30DV25 MILYUFC30HV25 MILYUFC30GV0S MILYUFC30DV0S MILYUFC30SV00 MILYUFC30HVIS
Filtry odśrodkowe, membrana mikroporowata Ultrafree®-MC
Filtry Filtry i koncentratory wirówkowe
Ultrafree®-MC centrifugal devices are single-use, disposable PP filters used for removing particulates from aqueous biological solutions. Devices are available with a range of microporous membranes for fast filtration and highly reproducible performance.
  • Fast filtration and highly reproducible performance
Applications include clarification of aqueous and some solvent based samples, protein and nucleic acid sample preparation.