Drukuj
Wirówkowe jednostki filtracyjne z membraną mikroporową, Ultrafree-CL
Dostawca: MILLIPORE
UFC40GV00 UFC40HV00 UFC40VV00 UFC40VV25 UFC40GV25 UFC40DV25 UFC40SV25 UFC40HV25
MILYUFC40GV00EA 2410 PLN
MILYUFC40GV00 MILYUFC40HV00 MILYUFC40VV00 MILYUFC40VV25 MILYUFC40GV25 MILYUFC40DV25 MILYUFC40SV25 MILYUFC40HV25
Wirówkowe jednostki filtracyjne z membraną mikroporową, Ultrafree-CL
Filtry Filtry i koncentratory wirówkowe
These centrifugal filter units can be used for a number of applications, including removal of particles and precipitates from aqueous and some solvent-based samples. Ideal for use with protein and nucleic acid solutions.
  • Pre-sterilised units also available
  • Fast filtration and highly reproducible performance
  • Use in fixed-angle rotors for 15 ml tubes
Hydrophilic PVDF membranes and PP housing.