Drukuj
Filtry membranowe, PORAFIL® PE
Dostawca: MACHEREY-NAGEL
671020013 671020025 671020037 671020047 671020050 671040013 671040025 671040037 671040047 671040050 671100013 671100025 671100037 671100047 671100050 671500013 671500025 671500037 671500047 671500050
MANA671020013EA 1140 PLN
MANA671020013 MANA671020025 MANA671020037 MANA671020047 MANA671020050 MANA671040013 MANA671040025 MANA671040037 MANA671040047 MANA671040050 MANA671100013 MANA671100025 MANA671100037 MANA671100047 MANA671100050 MANA671500013 MANA671500025 MANA671500037 MANA671500047 MANA671500050
Filtry membranowe, PORAFIL® PE
Filtry Filtry membranowe
Polyester membranes are particularly suited for fine filtration, dust analysis, aerosol analysis and ultra-purification of solvents.
  • Hydrophilic