Risocaine (propyl-4-aminobenzoate) 98%
Dostawca: Thermo Fisher Scientific
L09066.18
L09066.09
L09066.0B
L09066.36
L09066.18EA
866
PLN
L09066.18
L09066.09
L09066.0B
L09066.36
Risocaine (propyl-4-aminobenzoate) 98%
Risocaine (propyl-4-aminobenzoate)
Purity: 98%
CAS No.: 94-12-2
Molecular Formula: C10H13NO2
Formula Weight: 179.22
Melting Point: 73-75°
Beilstein Reference: 1211203
Formuła:
C₁₀H₁₃NO₂ Temp. topnienia: 73…75 °C Temperatura przechowywania: Temperatura otoczenia |
Numer MDL:
MFCD00017111 Numer CAS: 94-12-2 Numer WE (dawny EINECS): 202-306-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...