Drukuj
Wkłady ekstrakcji do fazy stałej, Bond Elut™ LMS
Dostawca: VARIAN
12105021 12105023 12105024 12105025 12105026 12255021
551-0337EA 1460 PLN
551-0337 551-0339 551-0340 551-0341 551-0342 551-0404
Wkłady ekstrakcji do fazy stałej, Bond Elut™ LMS
Kolumny chromatograficzne
Bond Elut™ LMS polymeric sorbent lets you elute without having to add amine modifiers, buffers, or acids. The elimination of secondary interactions means that elution of analytes can be achieved with pure organic solvents or HPLC mobile phase-compatible solvent mixtures of low ionic strength. These characteristics are crucial to allow compatibility with LC/MS or other delicate analytical techniques.
  • Ultra clean styrene-divinylbenzene polymer
  • Optimised 75 μm particle size for reproducible flow
  • High capacity and surface area for efficient extraction