Drukuj
Strzykawki do autosamplera Blue line do 7693A ALS, z tłokami zakończonymi PTFE
Dostawca: Agilent
G4513-80203 G4513-80219 G4513-80210 G4513-80220 G4513-80208 G4513-80241 G4513-80228 G4513-80223 G4513-80222 G4513-80221 G4513-80233
HEWLG4513-80203EA 684 PLN
HEWLG4513-80203 HEWLG4513-80219 HEWLG4513-80210 HEWLG4513-80220 HEWLG4513-80208 HEWLG4513-80241 HEWLG4513-80228 HEWLG4513-80223 HEWLG4513-80222 HEWLG4513-80221 HEWLG4513-80233
Strzykawki do autosamplera Blue line do 7693A ALS, z tłokami zakończonymi PTFE
Strzykawki i igły Strzykawki do chromatografii
Autosampler syringes with fitted plunger are recommended for homogenous liquid samples.
  • Plungers are not interchangeable or replaceable
  • Fitted syringe is individually matched with the plunger for precision injection
These syringes are suitable for gas and liquid samples and help to reduce carry-over and increase syringe lifetime.