Drukuj
disodu winian dihydrat, pure
Dostawca: CHEMPUR
Synonim: Sodium L-(+)-tartrate dihydrate, L(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Sodium tartrate dibasic dihydrate, Disodium L-(+)-Tartrate Dihydrate, (R,R)-(+)-Disodium tartrate dihydrate, L(+)-Tartaric acid, disodium salt, dihydrate, Sodium L-tartrate dibasic dihydrate, L(+)-Sodium tartrate dibasic dihydrate, Sodium Tartrate, Dihydrate, (+)-Disodium L-tartrate dihydrate, (+)-Disodium-L-tartrat dihydrate
428106409.0250 428106409.0500 428106409.1000
CHMP428106409.0250EA 111 PLN
CHMP428106409.0250 CHMP428106409.0500 CHMP428106409.1000
disodu winian dihydrat, pure
disodu winian dihydrat
Formuła: NaOOC(CHOH)₂COONa·2H₂O
MW: 230.08 g/mol
Temp. topnienia: 57
Gęstość: 1.818 g/cm³ (20 °C)
Numer MDL: MFCD00150035
Numer CAS: 6106-24-7
Numer WE (dawny EINECS): 212-773-3
Klasyfikacja Merck : 12,08833
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39a75038f4ff5e6c234267b15540823ae361bed4&vid=89289572c48b0c6415f8f11e916a51f66e129474&prid=web&clid=SAPDAM