Drukuj
Zakręcane fiolki i zamknięcia, 13 mm
Dostawca: Agilent
5182-0551 5183-4448 5183-4450
HEWL5182-0551EA 289 PLN
HEWL5182-0551 HEWL5183-4448 HEWL5183-4450
Zakręcane fiolki i zamknięcia, 13 mm
Vialki
These screw top vials with precision-formed screw threads of 13-425 are ideal for consistent secure seal and can be used as wash, waste, or storage vials.