Benzohydroxamic acid ≥98%
Dostawca: Thermo Fisher Scientific
|
Uwaga
|
Synonim:
Benzhydroxamic acid, N-Hydroxybenzamide
A10509.09
A10509.14
A10509.09EA
620
PLN
A10509.09
A10509.14
Benzohydroxamic acid ≥98%
Benzohydroxamic acid
Purity: 98%
CAS No.: 495-18-1
Molecular Formula: C7H7NO2
Formula Weight: 137.14
Storage Temperature: 2°C to 8°C
Melting Point: ca 125° dec.
Beilstein Reference: 1907585
MDL No.: MFCD00002109
Formuła:
C₇H₇NO₂ |
Numer MDL:
MFCD00002109 Numer CAS: 495-18-1 Numer WE (dawny EINECS): 207-797-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...