Drukuj
Jednorazowe skalpele, Minor
Dostawca: SWANN MORTON
2381 2373 2382 2385 2386 2383 2371 2372 2375 2376
SWAN2381EA 116 PLN
SWAN2381 SWAN2373 SWAN2382 SWAN2385 SWAN2386 SWAN2383 SWAN2371 SWAN2372 SWAN2375 SWAN2376
Jednorazowe skalpele, Minor
Ostrza, skalpele i uchwyty do ostrzy Skalpele
These non sterile disposable scalpels consist of a stainless steel blade and a PS handle.
  • 'Thumb' scalpels with a protective guard
  • Ideal for surgical applications
Please note: The protective guard is NOT compatible with a standard metal handle and blade.
Informacje do zamówienia: Available in packets of 10 or bulk boxes of 100.