Drukuj
Tabletki 3,3'-Diaminobenzydyny (DAB) do wizualizacji peroksydazy, SIGMAFAST™, Sigma-Aldrich®
Dostawca: Merck
D4293-50SET D4293-5SET D4418-50SET D4418-5SET
SIALD4293-50SETEA 1180 PLN
SIALD4293-50SET SIALD4293-5SET SIALD4418-50SET SIALD4418-5SET
Tabletki 3,3'-Diaminobenzydyny (DAB) do wizualizacji peroksydazy, SIGMAFAST™, Sigma-Aldrich®
Zestawy do testów środowiskowych
Developed for use in immunohistology and immunoblotting as a precipitating substrate for the localization of peroxidase activity. DAB is the immunohistology substrate of choice as it produces an intense brown-black stain which is resistant to alcohol. Slides stained with DAB may be dehydrated, mounted in resinous media, and stored for future reference.
SIGMAFAST™ DAB Tablets require no additional buffers or steps to prepare an active substrate solution.
Each tablet set dissolved in deionized water yields a ready-to-use buffered solution containing DAB and urea hydrogen peroxide.
Sposób pakowania: Tablets are individually packaged in foil packets.
Uwaga: Hazardous - please see SDS for details. Store the tablets at –20 °C.