Drukuj
Koszula, długi rękaw, Match 3-function
Dostawca: FRISTADS KANSAS
1004689402XL 1004689403XL 1004689404XL 100468940L 100468940M 100468940S 100468940XL 100468940XS
113-5349EA 235 PLN
113-5349 KWIN1004689403XL KWIN1004689404XL 113-5350 113-5351 113-5352 113-5348 KWIN100468940XS
Koszula, długi rękaw, Match 3-function
Odzież robocza
These functional shirts made of 58% polyester and 42% cotton provide protection against the cold. The insulating 3-function fabric ensures that moisture is absorbed quickly and the body remains dry and warm.
  • Crew neck
  • Long-sleeved
  • Lengthened back
Weight: 205 g/m²