Drukuj
Kamizelki, Fristads® PR54-521 Wzór A, ciemnoszary
Dostawca: FRISTADS KANSAS
8509411312XL 850941131L 850941131M 850941131S 850941131XL 850941131XS 8509412102XL 850941210L 850941210M 850941210S 850941210XL 850941210XS 8509412312XL 850941231L 850941231M 850941231S 850941231XL 850941231XS 8509413312XL 850941331L 850941331M 850941331S 850941331XL 850941331XS 8509414402XL 850941440L 850941440M 850941440S 850941440XL 850941440XS 8509415202XL 850941520L 850941520M 850941520S 850941520XL 850941520XS 8509415302XL 850941530L 850941530M 850941530S 850941530XL 850941530XS 8509415392XL 850941539L 850941539M 850941539S 850941539XL 850941539XS 8509416402XL 850941640L 850941640M 850941640S 850941640XL 850941640XS 8509417302XL 850941730L 850941730M 850941730S 850941730XL 850941730XS 8509419402XL 850941940L 850941940M 850941940S 850941940XL 850941940XS 8509419412XL 850941941L 850941941M 850941941S 850941941XL 850941941XS
113-5214EA 0 PLN
113-5214 113-03615 113-03616 113-03617 113-5213 113-03618 113-5210 113-03607 113-03608 113-03609 113-5209 113-03610 113-5216 113-03619 113-03620 113-03621 113-5215 113-03622 113-5218 113-03623 113-03624 113-03625 113-5217 113-03626 113-5220 113-03627 113-03628 113-03629 113-5219 113-03630 113-5200 113-03587 113-03588 113-03589 113-5199 113-03590 113-5204 113-03595 113-03596 113-03597 113-5203 113-03598 113-5202 113-03591 113-03592 113-03593 113-5201 113-03594 113-5206 113-03599 113-03600 113-03601 113-5205 113-03602 113-5208 113-03603 113-03604 113-03605 113-5207 113-03606 113-5222 113-03631 113-03632 113-03633 113-5221 113-03634 113-5212 113-03611 113-03612 113-03613 113-5211 113-03614
Kamizelki, Fristads® PR54-521 Wzór A, ciemnoszary
Odzież robocza
Functional vests made from 65% polyester and 35% cotton; with napped inner.
  • Zip fastening
  • 2 breast pockets with flap; front pockets; inner pocket
  • Back pleats; waistband adjustable using press studs, plastic press studs
  • Colour contrasting: Upper collar, bottom edge of zip cover and pocket edges
Material weight: 260 g/m²
Basic colour: Dark grey