Drukuj
Kaski zabezpieczające, 1125
Dostawca: CENTURION SAFETY PRODUCTS
S03EWF S03EGF S03EHVOA S03EHVYA S03EOA S03EOF S03EWA S03EBA S03EYF S03ERF S03EGA S03EBF S03BA S03GA S03OA S03RA S03WA S03YA
111-1520EA 75.4 PLN
111-1520 111-1515 111-1516 111-1517 111-1297 111-1518 111-1299 111-1294 111-1521 111-1519 111-1295 111-1514 111-1260 111-1301 111-1303 111-1304 111-1305 111-1306
Kaski zabezpieczające, 1125
Ochrona głowy Kaski ochronne
Impact resistant protective helmet with helmet shell made from high density polyethylene (HDPE). Suitable for the construction industry.
  • With six point terylene inner cradle
  • Front sweatband made from brushed nylon fabric
  • Ear protection, face protection shield and chin strap available as accessories
Size: 510 to 630 mm.
Weight: 365 g.
EN 397 (unvented version); Electrical Resistance and Molten Metal applicable to unvented shells only.
Certyfikaty: EN 397 (unvented version); Electrical Resistance and Molten Metal applicable to unvented shells only.