Drukuj
Mieszadełka magnetyczne, PTFE, ośmiokątne, różne, Spinpak®, Bel-Art
Dostawca: Bel-Art Products, a Part of SP
F371630000 F371580010
BELAF371630000EA 573 PLN
BELAF371630000 BELAF371580010
Mieszadełka magnetyczne, PTFE, ośmiokątne, różne, Spinpak®, Bel-Art
Mieszadła Mieszadełka magnetyczne
Eight-sided Spinbar® octagonal magnetic stirring bars provide greater surface area and added turbulence when compared to smooth cylindrical bars.
  • Integral pivot ring on each stirring bar aids in reducing friction and chattering
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE low friction coating, durable and inert
Spinbar® Restrainer fits comfortably in one hand and retains Spinbar® magnetic stirring bars up to 80 mm (3") long by holding them against vessel walls; eliminates splashing hazards when pouring.
Certyfikaty: SGS certified.
Informacje do zamówienia: The set includes a restrainer and various Spinbar® of different sizes: ½ x ⅛", ½ x ⁵/₁₆", 1 x ⁵/₁₆" 1½ x ⁵/₁₆", 2 x ⁵/₁₆" .