Methyl-2-aminothiophene-3-carboxylate 97%
Dostawca: Thermo Fisher Scientific
Synonim:
Methyl 2-aminothiophene-3-carboxylate
H55482.03
H55482.06
H55482.03EA
247
PLN
H55482.03
H55482.06
Methyl-2-aminothiophene-3-carboxylate 97%
Methyl-2-aminothiophene-3-carboxylate
Purity: 97%
CAS No.: 4651-81-4
Molecular Formula: C6H7NO2S
Formula Weight: 157.19
Melting Point: 76-81°
MDL No.: MFCD00159547
Formuła:
C₆H₇NO₂S MW: 157.19 g/mol |
Numer MDL:
MFCD00159547 Numer CAS: 4651-81-4 |