Drukuj

Glycylglycine (Gly-Gly)

Dostawca: BIOMOL RESEARCH LABORATORIES

05008-1 05008-2
BMOL05008-1EA 0 PLN
BMOL05008-1 BMOL05008-2
Glycylglycine (Gly-Gly)
Białka

  • EINECS: 209-127-8
  • GHS symbol and signal word: GHS07 and warning

Zobacz ceny


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR