Drukuj
Kuwety jednorazowe
Dostawca: KARTELL
1937 1938 1939 1940 1941 1948 1960 1961
634-8532EA 106 PLN
634-8532 634-8535 634-8536 634-0326 634-8533 634-8537 634-8531 634-8530
Kuwety jednorazowe
Kuwety Kuwety spektrofotometryczne
Disposable cuvettes made from optical polystyrene (PS) or PMMA with a transmission value at 300 nm of 70%.
  • 44,5 to 45 mm height