Drukuj

wodoroortofosforan (V) disodu dihydrat, pure

Dostawca: CHEMPUR

Synonim: sec-Sodium phosphate, Sodium monohydrogen phosphate dihydrate, Sodium hydrogenphosphate dihydrate, di-Sodium hydrogen orthophosphate dihydrate, Sodium phosphate dibasic dihydrate, Disodium phosphate, di-Sodium hydrogen phosphate dihydrate, Sorensen's salt

427992808.5000 427992808.0500 427992808.1000 427992808.9010
CHMP427992808.5000EA 462 PLN
CHMP427992808.5000 CHMP427992808.0500 CHMP427992808.1000 CHMP427992808.9010
wodoroortofosforan (V) disodu dihydrat, pure
wodoroortofosforan (V) disodu dihydrat
Formuła: Na₂HPO₄·2H₂O
Gęstość: 2,1 g/cm³ (20 °C)
Temperatura przechowywania: Temperatura otoczenia
Numer MDL: MFCD00149182
Numer CAS: 10028-24-7
Numer WE (dawny EINECS): 231-448-7

Zobacz ceny


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR