Drukuj

wodoroortofosforan (V) disodu dihydrat, pure analytical reagent

Dostawca: CHEMPUR

Synonim: sec-Sodium phosphate, Sodium monohydrogen phosphate dihydrate, Sodium hydrogenphosphate dihydrate, di-Sodium hydrogen orthophosphate dihydrate, Sodium phosphate dibasic dihydrate, Disodium phosphate, di-Sodium hydrogen phosphate dihydrate, Sorensen's salt

117992801.0050 117992801.0100 117992801.0250 117992801.0500 117992801.1000 117992801.9010
CHMP117992801.0050EA 54.1 PLN
CHMP117992801.0050 CHMP117992801.0100 CHMP117992801.0250 CHMP117992801.0500 CHMP117992801.1000 CHMP117992801.9010
wodoroortofosforan (V) disodu dihydrat, pure analytical reagent
wodoroortofosforan (V) disodu dihydrat
Formuła: Na₂HPO₄·2H₂O
MW: 177.99 g/mol
Temp. topnienia: 92.5
Gęstość: 2.1 g/cm³ (20 °C)
Temperatura przechowywania: Temperatura otoczenia
Numer MDL: MFCD00149182
Numer CAS: 10028-24-7
Numer WE (dawny EINECS): 231-448-7

Zobacz ceny


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR