Drukuj

Octan manganu(II) tetrahydrat, pure

Dostawca: CHEMPUR

Synonim: Manganese acetate tetrahydrate, Manganese diacetate tetrahydrate, Acetic acid manganese (II) salt tetrahydrate

426167508.0500 426167508.1000 426167508.9010
CHMP426167508.0500EA 76.4 PLN
CHMP426167508.0500 CHMP426167508.1000 CHMP426167508.9010
Octan manganu(II) tetrahydrat, pure
Octan manganu(II) tetrahydrat
Formuła: (H₃CCOO)₂Mn·4H₂O
MW: 245.09 g/mol
Temp. topnienia: 300
Gęstość: 1.58 g/cm³ (20 °C)
Numer MDL: MFCD00062552
Numer CAS: 6156-78-1
Numer WE (dawny EINECS): 211-334-3
Klasyfikacja Merck : 13,05746

Zobacz ceny


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR