Drukuj
Butelki z szeroką szyjką, z zakrętką
Dostawca: LAMAPLAST
413 412 411 410 409 408
215-1364EA 53.5 PLN
215-1364 215-0914 215-0913 215-0912 215-0911 215-0910
Butelki z szeroką szyjką, z zakrętką
Butelki i butle Butle plastikowe
LDPE, translucent.
  • Good chemical resistance, heat resistant up to 70 °C
  • Leakproof
  • Graduated
  • Suitable for food contact
Double closure cap, shoulder and perforated cap to allow the installation of seals and labels to ensure the integrity of the contents. Can be used anywhere in the laboratory and for sampling. Wide mouth for rapid filling and dispensing of powders, pastes and liquids.