Drukuj
Nośniki do wody pitnej/badanie wody
Dostawca: Merck
1.07680.0500 1.11471.0500
1.07680.0500EA 695 PLN
1.07680.0500 1.11471.0500
Nośniki do wody pitnej/badanie wody
Media do mikrobiologii Podłoża sypkie do mikrobiologii
Clean, fresh water is one of the most important issues facing humanity today. Also in the food and pharma industries the examination of water is prescribed. Granulated media from Merck are produced under strict guidelines and quality is controlled according to ISO 11133.
  • All your requirements - a wide range of dehydrated media or ready to use media such as Merckoplate® together with suitable filtering systems
  • High quality products at economical prices
  • All media comply with ISO standards
Ask for our Water Brochure - Microbiological examination Merck W 286135. GN-6 Cellulose filters from Pall are recommended for the examination of drinking water.