Drukuj
Wielokanałowe pipety, automatyczne, Finnpipette™ F2
Dostawca: Thermo Fisher Scientific
4662000 4662010 4662020 4662030 4662040 4662050 4662060 4662070 4662080 4662090
613-2720EA 5050 PLN
613-2720 613-2721 613-2722 613-2723 613-2724 613-2725 613-2726 613-2727 613-2728 613-2729
Wielokanałowe pipety, automatyczne, Finnpipette™ F2
Pipety Pipety i dozowniki seryjne
Finnpipette™ F2 offers superior robustness and durability even in the most demanding applications. Available as 8-, 12- and 16-channel models making it the ideal choice for both 96 and 384 microplate applications.
  • Fully autoclavable - No need to dismantle before autoclaving
  • Made of high-quality PVDF that withstands exposure to chemicals and UV light
  • Advanced Volume Gearing mechanism (AVG) – self-supporting modular mechanism increases accuracy, precision and durability, with light force and smaller increments for volume adjustment
  • Large, contrasted Ergovisio display with white numbers on black background for increased volume visibility and security
  • Soft-touch tip ejection to eject the tips with very light force
  • Superior handle ergonomics and very low force pipetting movement
  • Super blow-out piston for volumes of 50 µl and below, which ensures the delivery of micro-size drops
Informacje do zamówienia: Supplied with opening and calibration key, maintenance set (incl. grease and O-rings), calibration report, 3 year warranty certificate (extended to 5 years with web registration) and user's guide (English, French, German and Spanish).