Drukuj
Pojemnik próbek CellStor Pot™
Dostawca: CELLPATH
BAF-6100-08A BAF-9000-08A BAF-9100-08A BAF-6000-08A
720-2323EA 0 PLN
720-2323 720-2324 720-2325 CLLPBAF-6000-08A
Pojemnik próbek CellStor Pot™
Pojemniki na próbki Pojemniki na próbki
Prefilled specimen containers.
  • Leak-proof lid for safe transportation and specimen storage
  • With traceable badge number, safety data sheet in the packaging