Drukuj

winian diamonu, pure zA

Dostawca: CHEMPUR

Synonim: (R,R)-(+)-Diammonium tartrate, (R,R)-(+)-Ammonium tartrate dibasic, Ammonium tartrate dibasic, L(+)-Tartaric acid diammonium salt, (+)-Diammonium L-tartrate, (R,R)-(+)-Tartaric acid diammonium salt, L(+)-Ammonium tartrate dibasic, Ammonium L-(+)-tartrate

111404507.0100 111404507.0250 111404507.0500 111404507.1000
CHMP111404507.0100EA 89.7 PLN
CHMP111404507.0100 CHMP111404507.0250 CHMP111404507.0500 CHMP111404507.1000
winian diamonu, pure zA
winian diamonu
Formuła: C₄H₁₂N₂O₆
MW: 184.15 g/mol
Gęstość: ∼1.601 g/cm³ (20 °C)
Temperatura przechowywania: Temperatura otoczenia
Numer MDL: MFCD00013073
Numer CAS: 3164-29-2
Numer WE (dawny EINECS): 221-618-9

Zobacz ceny


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR