Drukuj
Butelki do bezpiecznego mycia, szeroka szyjka, z nadrukowaną nazwą, Bel-Art
Dostawca: Bel-Art Products, a Part of SP
F116460628 F116460459 F116460620 F116460622 F116460623 F116460624 F116460638 F116466032 F116461132 F116460621 F116460611 F116460612 F116460614 F116460627 F116460631 F116460639 F116462232 F116462432 F116463832 F116466250 F116460640
215-6233EA 470 PLN
215-6233 BELAF116460459 BELAF116460620 BELAF116460622 BELAF116460623 BELAF116460624 BELAF116460638 BELAF116466032 BELAF116461132 BELAF116460621 BELAF116460611 BELAF116460612 BELAF116460614 BELAF116460627 BELAF116460631 BELAF116460639 BELAF116462232 BELAF116462432 BELAF116463832 BELAF116466250 BELAF116460640
Butelki do bezpiecznego mycia, szeroka szyjka, z nadrukowaną nazwą, Bel-Art
Butelki i butle Tryskawki
LDPE, with PP spray cap.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Wide opening for easy adn safe filling
These bottles include a 53 mm wide PP closure with leakproof design for dispensing most common solvents. Easy-to-read, LDPE solvent-resistant, printed bottles include: Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information, making your workplace a safer place. Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200.