Drukuj

Karbazyd metylu, Sigma-Aldrich®

Dostawca: Merck

Synonim: Methyl hydrazinecarboxylate, Methyl hydrazinocarboxylate

M-Clarity™ quality level = MQ200

151653-100G 151653-500G
SIAL151653-100GEA 318 PLN
SIAL151653-100G SIAL151653-500G
Karbazyd metylu, Sigma-Aldrich®
Karbazyd metylu
Formuła: C₂H₆N₂O₂
MW: 90.08 g/mol
Numer MDL: MFCD00007594
Numer CAS: 6294-89-9
Numer WE (dawny EINECS): 228-560-3
Jednostka: 2811
ADR: 6.1 III

Zobacz ceny


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR