Drukuj
Złączki rurowe z uziemieniem, do ciśnienia do 5 barów, BOLA
Dostawca: Bohlender
H1001-04 H1001-14 H1001-12 H1001-10 H1001-06
BOHLH1001-04EA 169 PLN
BOHLH1001-04 BOHLH1001-14 BOHLH1001-12 BOHLH1001-10 BOHLH1001-06
Złączki rurowe z uziemieniem, do ciśnienia do 5 barów, BOLA
Adaptery i złącza Adaptery i złącza do węży
PTFE, white. For connecting tubes or tubing to vessels with ground joint. For inserting and fixing probes, thermometers, dip tubes or cables.
  • Temperature resistance: −200 to +250 °C
  • Universal chemical resistance
  • Autoclavable
  • Vacuum: suitable
Fitting made of PTFE for transition from ground joints to metric threads for connecting hard-walled tubing (e.g. PTFE, PFA or FEP). With nuts made of glass-fibre reinforced PTFE, body with rings and knurled grip for opening. The product is only exposed to PTFE.