Drukuj
Systemy oczyszczania wody, RiOs™ 30/50/100/150/200
Dostawca: Merck
ZR0S50200SAVT ZR0S50150SAVT ZR0S50100SAVT ZR0S50030SAVT ZR0S50050SAVT
MILLZR0S50200SAVTEA 0 PLN
MILLZR0S50200SAVT MILLZR0S50150SAVT MILLZR0S50100SAVT MILLZR0S50030SAVT MILLZR0S50050SAVT
Systemy oczyszczania wody, RiOs™ 30/50/100/150/200
Systemy oczyszczania wody Systemy czystej i ultra czystej wody
RiOs™ systems are used to produce large volumes of Type 3 water from potable tap water. The RiOs™ system incorporates both the complete water purification sequence for the production of laboratory-grade water, as well as the entire control and monitoring functions for the total water purification system. Flow rate and water quality can be upgraded in RiOs™ systems.
  • Provides up to 8 000 L of RO water per day
  • Built-in UV lamp (254 nm) for bacteria-sensitive applications
  • Easy-to-replace pretreatment packs, tailored to the feed water source
  • Built-in