Drukuj
Zestawy pompy beczkowej do stężonych kwasów i zasad, MA II
Dostawca: LUTZ PUMPEN
0205-202 0205-211 0205-212 0205-201
223-0089EA 18610 PLN
223-0089 223-0090 223-0091 223-0088
Zestawy pompy beczkowej do stężonych kwasów i zasad, MA II
Pompy Pompy do beczek
This pump set is specially adapted for pumping concentrated acids and alkalis such as chloric acid, chromic acid, sulphuric acid, nitric acid, hydrofluoric acid and sodium hypochlorite. Pump tube and nozzle in PVDF and drum adapter made from PP.
  • Fast assembly, immediately ready for action
  • Versatile use
  • Interchangeable motors and pump tubes
  • Modular construction
  • Easy maintenance and service
The pump set consists of a motor MA II 3 or MA II 5, pump tube in polyvinylidenefluoride (PVDF) with Hastelloy-C shaft, 2,5 m special chemical hose 3/4" and hose connections, nozzle in PVDF, hose clips, drum adapter in PP.