Drukuj
Papier samoprzylepny do pomieszczeń czystych, TexBond® 20
Dostawca: Texwipe
TX5719 TX5711_U TX5728 TX5731 TX5729 TX5723 TX5724 TX5711
818-0270EA 99.3 PLN
818-0270 TEXWTX5711_U 818-0273 818-0275 818-0274 818-0271 818-0272 818-0269
Papier samoprzylepny do pomieszczeń czystych, TexBond® 20
Artykuły papiernicze
TexBond® is a cleanroom bond paper with exceptional contamination control characteristics needed in critical environments. It has been especially formulated to address the needs of users requiring a paper that retains low ion and particle levels with significantly improved copying and writing qualities. This paper will yield sharp copies that will not show through to the next sheet due to its high opacity rating.
  • Exceptional copying ability
  • Laser printer and photocopier compatible
  • Clear reproductions
  • Autoclavable for use in sterile environments
  • Weight: 75 g/m²