Drukuj
Zintegrowane zawieszane pokrywy głowic, Versaflo™ serii S
Dostawca: 3M
S657 S333LG S333SG S533L S533S S133L S133S S757 S655E
111-1118EA 949 PLN
111-1118 111-1110 111-1111 111-1112 111-1113 111-1108 111-1109 111-1119 111-2348
Zintegrowane zawieszane pokrywy głowic, Versaflo™ serii S
Maski oddechowe Systemy Oddechowe
The 3M™ S-series headcovers offer head and face protection, as well as providing eye and face protection from liquid splash and low energy impact (grade F). They are suitable for applications that require frequent replacement of the entire headgear assembly, such as pharmaceutical manufacturing and laboratory work, or in situations where there is less soiling of the outer fabric. All headcovers can be used in conjunction with 3M Powered and Supplied Air systems (e.g. Jupiter™ and Dustmaster™), thus offering respiratory protection.
  • Improved shape and visibility
  • Excellent airflow
  • Comfortable to wear for long periods thanks to loose-fitting design
  • Nominal protection factor: 500
S-133: Disposable headcover for general purpose applications
S-333G: Disposable grey headcover with increased chemical and scratch resistance, it is therefore suitable for applications in the chemical industry. The material is soft, quiet, durable and low linting.
S-533: Disposable headcover with hood made of soft low linting fabric that readily drapes over the user and provides additional neck and shoulder coverage. The headcover features increased chemical and scratch resistance.
S-655: Reusable Premium headcover with removable head harness and connector allowing these low wear parts to be transferred into a replacement hood. Combines face (low energy impact and liquid splash), neck, shoulder and respiratory protection.
S-657: Reusable Premium headcover with hood that features a double shroud design for its respiratory seal.
S-757: Reusable Premium headcover with hood which features a fabric that is specifically intended to help capture paint overspray.
S-333G: Grey headcover with increased chemical and scratch resistance, it is therefore suitable for applications in the chemical industry. The material is soft, quiet, durable and low-linting.
S-433: Headcover with hood made of a general-purpose, cost effective material. The headcover features increased chemical and scratch resistance.
S-533: Headcover with hood made of soft low-linting fabric that readily drapes over the user and provides additional neck and shoulder coverage. The headcover features increased chemical and scratch resistance.
S-655: Headcover with hood for general purpose applications. The knitted inner collar is shorter and thinner than previous models.
Certyfikaty: EN 166; meets highest respiratory class 12941 TH3 when used with respiratory systems.
Uwaga: *One size fits all.