Drukuj
Kolumny SPE Retain™-CX HyperSep™
Dostawca: Thermo Fisher Scientific
60107-308 60107-301 60107-303 60107-304 60107-305 60107-306 60107-307 60107-311 60107-312 60107-314 60107-315
549-0647EA 468 PLN
549-0647 549-0640 549-0642 549-0643 549-0644 549-0645 549-0646 549-0648 549-0649 549-0650 549-0651
Kolumny SPE Retain™-CX HyperSep™
Kolumny chromatograficzne
HyperSep™ Retain™-CX polymeric SPE material is a high purity, highly porous, polystyrene DVB material partially functionalised with sulphonic acid groups to give balanced retention of basic and non polar analytes. HyperSep™ Retain™-CX is ideal for the retention of a wide range of drugs of abuse, including basic and neutral drugs.
  • High and consistent recoveries
  • Fast and easy sample preparation and method development
  • pH stable zero to 14
HyperSep™ Retain™-CX offers reproducible and reliable sample preparation through excellent  batch-to-batch reproducibility. Many of the problems associated with traditional silica-based materials, such as inconsistent recoveries due to sorbent drying, are eliminated when using HyperSep™ Retain™-CX. This can lead to fast sample preparation and method development.