Drukuj
Słyżkie złącze adaptery Luer , Upchurch Scientific®
Dostawca: UPCHURCH SCIENTIFIC
P-624 B-310 P-604 P-605 P-618 P-618L P-619 P-625 P-628 P-628S P-629 P-642 P-655 P-656 P-657 P-658 P-659 P-660 P-661 P-675 P-677 P-678 P-683 P-686
554-1792EA 51.8 PLN
554-1792 554-5774 554-5683 554-4725 554-1790 554-5684 554-1791 554-1793 554-1794 554-4726 554-4727 554-1795 554-1796 554-1797 554-3139 554-1798 554-1799 554-3140 554-3141 554-1800 554-3147 554-1801 554-3150 554-3151
Słyżkie złącze adaptery Luer , Upchurch Scientific®
Okucia i złącza
Upchurch Scientific® quick connect Luer adapters provide the opportunity to adapt either a female Luer or male Luer connection to a threaded connection. Conform to ISO requirements for medical Luer taper configuration and performance (45 psi/3,1 bar).