Drukuj
Zatyczki do uszu na pałąku, E-A-R™ Caboflex™
Dostawca: 3M
CS01000 CF01000
111-2200EA 2260 PLN
111-2200 111-1553
Zatyczki do uszu na pałąku, E-A-R™ Caboflex™
Zatyczki do uszu Zatyczki do uszu
This lightweight semi-aural hearing protector is ideal when moving in and out of high noise areas, fitting comfortably around the neck when not in use. The conical shaped pods can swivel to align with the ear canal, providing a move resistant seal against noise.
  • Flexible, lightweight, dielectric chin band won't interfere with hard hat or glasses
  • Slow recovery foam: Improved fit of pods in ear canal
  • Replacement pods are available for increased economy
SNR: 21 dB
Certyfikaty: EN 352-2