Drukuj
Taśmy pakowe, taśmy filamentowe, 895, Scotch®, 3M
Dostawca: 3M
8951950 8952550 8951250 8955050
819-1440EA 125 PLN
819-1440 819-1441 MMMA8951250 MMMA8955050
Taśmy pakowe, taśmy filamentowe, 895, Scotch®, 3M
Taśmy
These 895 filament packaging tapes are made of ScotchPro™ PP film with a synthetic rubber adhesive specifically formulated to provide balanced performance, having both high adhesion and long term holding power.
  • Reinforced along its length with continuous glass yarn filaments for very high tensile strength and excellent nick resistance
  • Suitable for hand held manual dispensers and semiautomatic L-Clip applicator
  • Application most easily accomplished at room temperature
Length: 50,0 m
Colour: Transparent