Drukuj
Szalki Petriego, jednorazowe, markowe
Dostawca: Brand
452000 452005
BRND452000EA 501 PLN
BRND452000 BRND452005
Szalki Petriego, jednorazowe, markowe
Szalki do hodowli Szalki Petriego
PS, transparent, with lid. Available with or without vent lid. Dishes and lids are assembled robotically to reduce the risk of contamination, in particular bacterial count.