Drukuj
Taśmy etykietujące, winylowe, ogólnego przeznaczenia
Dostawca: 3M
764B5033 764GE5033 764O5033 764R5033 764S5033 764T5033 764W5033 764I-GR 764I-GY
819-0110EA 1530 PLN
819-0110 819-0111 819-0112 819-0113 819-0115 819-0116 819-0117 819-0140 819-0139
Taśmy etykietujące, winylowe, ogólnego przeznaczenia
Taśmy
3M™ general purpose tapes are made of vinyl with rubber adhesive. They are used in non critical applications such as colour coding, bundling and safety marking, temporary marking of parts or machines, protect parts, equipment or products from abrasions.
  • Use for taping in all non-critical applications
  • Protects parts, equipment, and products from abrasions
  • Bundle items, bundle and temporarily mark parts, machines, plus much more
  • Maintains vivid colour over time for long-lasting quality
  • Can be applied by hand or with floor applicator
  • Ideal for marking applications in light traffic areas
  • The 4 mil (0,10 mm) PVC backing features a rubber adhesive
Find dozens of ways to use 3M™ general purpose vinyl tape 764. This vinyl tape is perfect for all non-critical applications. The tape can be used for a wide variety of applications, including protecting parts, equipment, and products from abrasions. It is non-corrosive, flexible, abrasion-resistant, conformable, wear-resistant, easy to apply, versatile, and moisture-resistant
Adhesion strength - range: 21 N/100 mm
Overall length: 33 m
Tensile strength: 228 N/100 mm