n-Heksatriakontan ≥97%
Dostawca: Thermo Fisher Scientific
31641.09
31641.18
31641.09EA
549
PLN
31641.09
31641.18
n-Heksatriakontan ≥97%
n-Heksatriakontan
Purity: 97+%
CAS No.: 630-06-8
Molecular Formula: CH3(CH2)34CH3
Formula Weight: 506.95
Physical Form: Wax
Density: 0.779
Refractive Index: 1.4573
Appearance: Off-white
Melting Point: 76.2°
Boiling Point: 497°
MDL No.: MFCD00009413
Formuła:
H₃C(CH₂)₃₄CH₃ Temp. Wrzenia: 497 °C Gęstość: 0,779 Temperatura przechowywania: Temperatura otoczenia |
Numer MDL:
MFCD00009413 Numer CAS: 630-06-8 Numer WE (dawny EINECS): 211-127-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...