3-Merkapto-2-pentanon 97% stabilised
Dostawca: Thermo Fisher Scientific
B21797.06
B21797.03
B21797.06EA
1180
PLN
B21797.06
B21797.03
3-Merkapto-2-pentanon 97% stabilised
3-Merkapto-2-pentanon
Purity: 98%
CAS No.: 67633-97-0
Molecular Formula: C5H10OS
Formula Weight: 118.19
Storage Temperature: -30°C to -10°C
Refractive Index: 1.4660
UN No.: 3336
MDL No.: MFCD02093290
Formuła:
C₅H₁₀OS |
Numer MDL:
MFCD02093290 Numer CAS: 67633-97-0 |