Drukuj
Stojaki na mikropłytki 96-dołkowe, BRAND
Dostawca: Brand
781565 781566
738-0137EA 3660 PLN
738-0137 738-0138
Stojaki na mikropłytki 96-dołkowe, BRAND
Statywy do probówek Tube Racks
Non sterile, suitable for use with robots
  • Tubes and racks (not cover mats or strips) are autoclavable at 121 °C, (2 bar), acc. DIN EN 285
  • Tubes, racks and lids can withstand temperatures as low as −80 °C
  • Coded tubes (A1 to H12) and rack sides suitable for barcode labels simplify sample organisation