Drukuj
Złączki do rurek, z zaworem, Nalgene®, Thermo Fisher Scientific
Dostawca: Thermo Fisher Scientific
6177-0250 6177-0375
NALG6177-0250EA 653 PLN
NALG6177-0250 NALG6177-0375
Złączki do rurek, z zaworem, Nalgene®, Thermo Fisher Scientific
Okucia i złącza
PP, beige.
  • Fluid flow stops when fitting halves are separated
  • Designed for one-handed quick disconnect with flexible tubing
  • Provides a leakproof† connection between two tubes, but may require clamps
Maintain the integrity of transfer liquids. The barbed fitting on each end and internal valve in the middle, which seals when the fittings are removed, are ideal for use in developing in-house fluid transfer systems.