Drukuj
Ściereczki do ogólnych zastosowań, WYPALL* X80, kodowane kolorami
Dostawca: KIMBERLY CLARK
7565 7566 7567 7568
115-1550EA 1420 PLN
115-1550 115-1551 115-1552 115-1553
Ściereczki do ogólnych zastosowań, WYPALL* X80, kodowane kolorami
Chusteczki Ściereczki wielorazowe Ściereczki uniwersalne
WYPALL* X80 wipes are made of the Long-lasting and strong HYDROKNIT* material, and therefore ideal for heavy-duty tasks Like wiping away grease, oil and cleaning up heavier soiling.
  • High absorbency coupled with great tear resistance
  • Can be used with water or solvents - just rinse out and re-use
  • Colour coded: easy to identify with specific tasks, reducing the risk of cross-contamination and confusion