Drukuj
Płytki do hodowli kolagenu IV, Corning® BioCoat™, Corning
Dostawca: Corning
354453 354416
734-0139EA 1280 PLN
734-0139 734-0122
Płytki do hodowli kolagenu IV, Corning® BioCoat™, Corning
Szalki do hodowli Szalki do hodowli komórkowych
PS coated with mouse collagen type IV. Collagen IV plays a role in the regulation of cell growth, differentiation and adhesion, as well as tissue formation.
Manufactured in a highly controlled environment and rigorously tested to assure product consistency and performance.