Drukuj
Łaźnie wodne, seria WB
Dostawca: P D UMWELTSCHUTZ UND PRUEFGERA
3297.1.00005 3296.1.00010 3296.1.00020
462-6718EA 0 PLN
462-6718 462-6606 462-6607
Łaźnie wodne, seria WB
Łaźnie Łaźnie wodne bez mieszania
This range of high grade steel water baths has a variety of accessories to provide versatile combinations and enable various different applications to be performed. The temperature can be displayed in Fahrenheit or Celsius, and it is possible to calibrate using the actual value. Baths have a screw cap outlet to enable the water to be emptied, a drain tap can be ordered separately if required. Baths can be used with test tube racks, clamps for Erlenmeyer flasks.
  • Easy operation via digital LED display
  • Microprocessor control with adjustable over and under temperature limits, audible and visual warning signals
  • Low liquid level protection to avoid any potential damage
  • Infinitely adjustable shaking frequency from 20 to 200 rpm, and shaking amplitude of 16 mm when using optional
Certyfikaty: Conform to DIN 12876-1 Class 1.
Informacje do zamówienia: Supplied without accessories, accessories must be ordered separately.