Drukuj
Pierścienie zaciskowe dla małych połączeń kołnierzowych
Dostawca: VACUUBRAND
20660000 20660001 20660002 20660003 20660010 20660011 20660012
185-1111EA 114 PLN
185-1111 185-1112 185-1113 185-1114 185-1121 185-1122 185-1123
Pierścienie zaciskowe dla małych połączeń kołnierzowych
Zaciski Zaciski uniwersalne
For securing and mechanical pressing of O-ring seals. Aluminium or stainless steel, non rusting.
  • Aluminium clamping rings with wing nut for fast connection without tools
  • Stainless steel clamping rings for assembly with allen screws; for applications in which corrosion is possible, requirements for low gas discharge and particularly clean conditions
  • System components for a wide range of connections